missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobaltous Chloride, Hexahydrate, ACS Grade, 98.0 to 102.0% , LabChem™

$279.46 - $279.46

Chemical Identifiers

CAS 7791-13-1
CAS Max % 100
Linear Formula CoCl2·6H2O
Formula Weight 237.92
UN Number UN3260
Chemical Name or Material Cobaltous Chloride, Hexahydrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC131901 Lab Chem
LC131901
500g Each for $279.46
Specifications Description & Specifications

Chemical Identifiers

7791-13-1
CoCl2·6H2O
UN3260
ACS Grade
100
237.92
Cobaltous Chloride, Hexahydrate

Specifications

237.92
1,924kg/m3
CoCl2·6H2O
7791-13-1
Hydrogen chloride
500g
86°C
1924kg/m3
Cobaltous Chloride, Hexahydrate
ACS Grade
Room Temperature
Crystals
Pink to red
Passes Test
Poly Bottle
Soluble in water
100
UN3260
98.0 to 102.0%, as CoCl2·6H2O

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing dust.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
If exposed or concerned: Get medical attention.
If swallowed: Rinse mouth.
Call a poison center/doctor if you feel unwell.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If experiencing respiratory symptoms: Call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

EINECSNumber : 231-589-4