Please login to your online account to display your discounted pricing

Cobaltous Chloride, For Nitrogen (Ammonia), Nitrogen (Organic), LabChem™

$80.54 - $80.54

Chemical Identifiers

Chemical Name or Material Cobaltous Chloride
CAS 7732-18-5
CAS Max % 94.46
CAS 7647-01-0
CAS Max % 4.34
CAS 7791-13-1
CAS Max % 1.2
Name Note For Nitrogen (Ammonia), Nitrogen (Organic)
Linear Formula CoCl2·6H2O
Formula Weight 237.92
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC132001

 
lab chem
LC132001
500mL Each for $80.54
Description & Specifications

Specifications

Formula Weight 237.92
Chemical Name or Material Cobaltous Chloride
CAS Max % 1.2
Decomposition Information Hydrogen chloride; Thermal decomposition generates: Corrosive vapors; Cobalt and compounds
Color Clear pink
Quantity 500mL
Name Note For Nitrogen (Ammonia), Nitrogen (Organic)
Physical Form Liquid
Linear Formula CoCl2·6H2O
CAS 7732-18-5
UN Number UN1789
Health Hazard 1 Danger
CAS Max % 94.46
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Harmful to aquatic life with long lasting effects.
Identification Passes Test
CAS 7647-01-0
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin: Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
CAS Max % 4.34
Solubility Soluble in water
CAS 7791-13-1
Storage Information Room Temperature
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin: Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Harmful to aquatic life with long lasting effects.