Please login to your online account to display your discounted pricing

Cobaltous Acetate Tetrahydrate (Crystalline/Certified), Fisher Chemical


Quantity 100g
Packaging Glass Bottle

Manufacturer: fisher scientific company  FLC364100

 View more versions of this product

Catalog No. C364-100

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 140°C
Purity Grade Certified Grade
Formula Weight 249.08
Quantity 100g
DOT Information DOT Class 9, : Miscellaneous
Synonym Acetic Acid Cobalt Salt
Nitrate 0.015% max.
Insoluble Matter 0.01% max.
pH 6.8
Physical Form Solid
Color Light Red
Sulfate 0.01% max.
ChemAlert Storage Symbol Gray
Health Hazard 2 WARNING!
Copper (Cu) 0.005% max.
Chloride 0.003% max.
Zinc (Zn) 0.03% max.
CAS 6147-53-1
Alkalis and Alkaline Earths 0.25% max.
Molecular Formula C4H6CoO4 · 4 H2O
UN Number 3077
Lead (Pb) 0.003% max.
Nickel (Ni) 0.15% max.
Identification Pass Test
Packaging Glass Bottle
Iron (Fe) 0.003% max.
Health Hazard 3 Emergency Overview
May cause cancer by inhalation. May impair fertility. May cause skin, eye, and respiratory tract irritation. May cause allergic respiratory and skin reaction. Possible risks of irreversible effects. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:3
Flammability:1
Instability:1
Linear Formula Co(CH3COO)2·4H2O
Chemical Name or Material Cobaltous Acetate Tetrahydrate
MDL Number MFCD00149569

Specifications

Chemical Name or Material Cobaltous Acetate Tetrahydrate
CAS 6147-53-1
Purity Grade Certified Grade
Physical Form Solid
Molecular Formula C4H6CoO4 · 4 H2O
Linear Formula Co(CH3COO)2·4H2O
Formula Weight 249.08
MDL Number MFCD00149569
Synonym Acetic Acid Cobalt Salt
Accessories
 

Emergency Overview
May cause cancer by inhalation. May impair fertility. May cause skin, eye, and respiratory tract irritation. May cause allergic respiratory and skin reaction. Possible risks of irreversible effects. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:3
Flammability:1
Instability:1

WARNING!

SDS & Safety Documents