Please login to your online account to display your discounted pricing

Cobaltocene, 98%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC405320100

 View more versions of this product

Catalog No. AC405320100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Metallic Crystals
Formula Weight 189.12
Color Black
Quantity 10g
Chemical Name or Material Cobaltocene
Assay Percent Range 98%
Infrared Spectrum Authentic
Molecular Formula C10H10Co
MDL Number MFCD00013749
CAS 1277-43-6
Packaging Glass bottle
Synonym Bis(cyclopentadienyl)cobalt
Health Hazard 1 GHS Signal Word: Warning
Elemental Analysis 97.5% min.
Health Hazard 2 GHS H Statement:
Flammable solid.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Cobaltocene
Assay Percent Range 98%
Molecular Formula C10H10Co
Formula Weight 189.12
MDL Number MFCD00013749
Synonym Bis(cyclopentadienyl)cobalt
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.

SDS & Safety Documents