Please login to your online account to display your discounted pricing

Cobaltocene, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  405320010

 View more versions of this product

Catalog No. AC405320010

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Black
Physical Form Metallic Crystals
Formula Weight 189.12
Quantity 1g
Elemental Analysis 97.5% min.
Chemical Name or Material Cobaltocene
CAS 1277-43-6
Assay Percent Range 98%
Molecular Formula C10H10Co
MDL Number MFCD00013749
Infrared Spectrum Authentic
Packaging Glass bottle
Synonym Bis(cyclopentadienyl)cobalt
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Flammable solid.

Specifications

Chemical Name or Material Cobaltocene
Assay Percent Range 98%
Molecular Formula C10H10Co
Formula Weight 189.12
MDL Number MFCD00013749
Synonym Bis(cyclopentadienyl)cobalt
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable solid.

SDS & Safety Documents