missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt wire, 2.0mm (0.08 in.) dia., Puratronic™, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 10945BS

 View more versions of this product

Catalog No. AA10945BS



Specifications

Specifications

Cobalt wire
10cm
7440-48-4
Co
atom
[Co]
Odorless
104730
Wire
Puratronic™
≈28.0g/m
2.0mm (0.08 in.) dia.
(metals basis)
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
58.933
CHEBI:27638
99.995%

Chemical Identifiers

7440-48-4
58.933
GUTLYIVDDKVIGB-UHFFFAOYSA-N
104730
cobalt
Co
MFCD00010935
atom
CHEBI:27638
[Co]
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents