missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt wire, 1.5mm (0.06 in.) dia., annealed, Puratronic™, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 44525BS

 View more versions of this product

Catalog No. AA44525BS



Specifications

Specifications

Cobalt wire
10cm
7440-48-4
Co
atom
[Co]
Odorless
104730
Wire, ≈15.8g/m
Puratronic™
≈15.8g/m
1.5mm (0.06 in.) dia., Annealed
(metals basis)
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
58.933
CHEBI:27638
99.995%

Chemical Identifiers

7440-48-4
58.933
GUTLYIVDDKVIGB-UHFFFAOYSA-N
104730
cobalt
Co
MFCD00010935
atom
CHEBI:27638
[Co]
Safety and Handling

Safety and Handling

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H350

EINECSNumber : 231-158-0

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents