Please login to your online account to display your discounted pricing

Cobalt tetramethoxyphenylporphyrin, 94%, ACROS Organics™

$255.35 - $255.35

Chemical Identifiers

Chemical Name or Material Cobalt tetramethoxyphenylporphyrin
CAS 28903-71-1
Assay ≥92%
Assay Percent Range 94%
Molecular Formula C48H36CoN4O4
Formula Weight 791.77
MDL Number MFCD00010724
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC405340250

  SDS
thermo fisher scientific
AC405340250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC405340010

  SDS
thermo fisher scientific
AC405340010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC405340050

  SDS
thermo fisher scientific
405340050
Glass bottle 5g Each for $255.35
Description & Specifications

Specifications

Assay ≥92%
Formula Weight 791.77
Physical Form Crystals or Powder
Quantity 25g
Color Purple to violet
CAS 28903-71-1
Chemical Name or Material Cobalt tetramethoxyphenylporphyrin
Assay Percent Range 94%
Molecular Formula C48H36CoN4O4
MDL Number MFCD00010724
Packaging Glass bottle
Infrared Spectrum Authentic