Please login to your online account to display your discounted pricing

Cobalt tetramethoxyphenylporphyrin, 94%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC405340010

 View more versions of this product

Catalog No. AC405340010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥92%
Physical Form Crystals or Powder
Formula Weight 791.77
Quantity 1g
Color Purple to violet
Chemical Name or Material Cobalt tetramethoxyphenylporphyrin
Assay Percent Range 94%
Molecular Formula C48H36CoN4O4
Infrared Spectrum Authentic
MDL Number MFCD00010724
CAS 28903-71-1
Packaging Glass bottle

Specifications

Chemical Name or Material Cobalt tetramethoxyphenylporphyrin
CAS 28903-71-1
Assay ≥92%
Assay Percent Range 94%
Molecular Formula C48H36CoN4O4
Formula Weight 791.77
MDL Number MFCD00010724