Please login to your online account to display your discounted pricing

Cobalt Sulfate, 7-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 125g
Packaging Poly Bottle

Manufacturer: avantor performance  169604

 View more versions of this product

Catalog No. 02002631

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 281.1
Physical Form Crystals
Chemical Name or Material Cobalt Sulfate
Molecular Formula CoSO4·7H2O
Packaging Poly Bottle
Name Note 7-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Quantity 125g
Specific Gravity 1L = 2.03kg
CAS 10026-24-1

Specifications

Chemical Name or Material Cobalt Sulfate
CAS 10026-24-1
Name Note 7-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula CoSO4·7H2O
Formula Weight 281.1