missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

$221.18 - $378.35

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
PLCO2-2Y
SDS View Documents Promotion Details
Spex Certiprep Llc
PLCO22Y
125mL Triple-leached LDPE bottle
Each for $221.18
 
PLCO22T
View Documents Promotion Details
Spex Certiprep Llc
PLCO22T
250mL Triple-leached LDPE bottle
Each for $314.32
 
PLCO2-2X
View Documents Promotion Details
Spex Certiprep Llc
PLCO22X
500mL Triple-leached LDPE bottle
Each for $378.35
 
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Co1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Identifiers

7440-48-4
58.93
GUTLYIVDDKVIGB-UHFFFAOYSA-N
CHEBI:27638
[Co]
Co
MFCD00010935
104730
cobalt

Specifications

7440-48-4,7732-18-5,7732-18-5
0.1,2.0,97.9
MFCD00010935
[Co]
58.93
CHEBI:27638
Triple-leached LDPE bottle
2% HNO3
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Cobalt Standard
0.1,2.0,97.9
Co
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
104730
Assurance
Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
125mL
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Safety and Handling

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Hazardous

Recommended Storage : Ambient

SDS
Product Certifications

Product intended for laboratory use only