Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis)

$817.35 - $817.35

Chemical Identifiers

Chemical Name or Material Cobalt sputtering target
CAS 7440-48-4
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00010935
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40836KS

  SDS
alfa aesar™
40836KS
1 each Each for $817.35
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Cobalt sputtering target
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD00010935
CAS 7440-48-4
Odor Odorless
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Quantity 1 each
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.