Please login to your online account to display your discounted pricing

Cobalt, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


For atomic absorption. (1mL = 1mg Co) Solute: Cobalt Nitrate Solvent: Dilute Nitric Acid

Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  SC193500

 View more versions of this product

Catalog No. SC193-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Boiling Point 100°C
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Color Light Red
pH 2
Vapor Pressure 17mmHg at 20°C
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Packaging Poly Bottle
CAS 7732-18-5
MDL Number MFCD00010935
Traceability to NIST LOT NUMBER USED COBALT REF. STD.
Chemical Name or Material Cobalt, Reference Standard Solution
Concentration 990 to 1010ppm
ChemAlert Storage Symbol White
CAS 7697-37-2
CAS 10026-22-9
Name Note 1000ppm ±1%

Specifications

Chemical Name or Material Cobalt, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 7697-37-2
CAS 10026-22-9
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00010935
Accessories
 

SDS & Safety Documents