missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt powder, 99.99% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer:  Alfa Aesar 1061014

 View more versions of this product

Catalog No. AA1061014



Specifications

Specifications

Cobalt powder
7440-48-4
Co
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
[Co]
58.933
CHEBI:27638
25g
99.99% (metals basis)
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
104730

Chemical Identifiers

7440-48-4
58.933
GUTLYIVDDKVIGB-UHFFFAOYSA-N
104730
cobalt
Co
MFCD00010935
kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
CHEBI:27638
[Co]
Safety and Handling

Safety and Handling

EINECSNumber : 231-158-0

Documents