missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt Powder, -400 mesh, 99.95% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4408530

 View more versions of this product

Catalog No. AA4408530



Specifications

Specifications

Cobalt
7440-48-4
99.95% (Metals basis)
MFCD00010935
atom
[Co]
Odorless
104730
Powder
250g
Black powder
Co
-400 Mesh
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
58.933
CHEBI:27638
Black

Chemical Identifiers

7440-48-4
58.933
GUTLYIVDDKVIGB-UHFFFAOYSA-N
104730
cobalt
Co
MFCD00010935
atom
CHEBI:27638
[Co]
Safety and Handling

Safety and Handling

P201-P202-P210-P240-P241-P260-P264b-P270-P271-P272-P280-P281-P284-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P363-P370+P378q-P501c

H228-H302-H317-H319-H330-H334-H335-H336-H350i-H360-H373-H500

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-158-0

RTECSNumber : GF8750000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents