missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt plasma standard solution, Alfa Aesar™, Specpure™

Manufacturer:  Alfa Aesar 13828AD

 View more versions of this product

Catalog No. AA13828AD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Cobalt
Clear liquid
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
104730
Liquid
Colorless
7440-48-4,7732-18-5,7732-18-5
Co
Nitric acid: 5.0%; Cobalt: 0.1%; Water: 94.9%
[Co]
58.933
CHEBI:27638
23 hPa (17mm Hg) at 20°C
50mL

Chemical Identifiers

7440-48-4
58.933
GUTLYIVDDKVIGB-UHFFFAOYSA-N
CHEBI:27638
[Co]
Co
MFCD00010935
104730
cobalt
Safety and Handling

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : Hazard Class: 8, Packing Group: III

Documents