missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt pieces, >99.9% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 10454A3

 View more versions of this product

Catalog No. AA10454A3



Specifications

Specifications

Cobalt
7440-48-4
99.9+% (Metals basis)
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
cobalt
58.933
CHEBI:27638
0.0000001 hPa
2kg
Gray pieces
Co
atom, co1+, cobalt, ion co1+, cobalto, cobaltum, hydride, hydrido, kobalt, moncation, powder
[Co]
Odorless
104730
Pieces
Gray

Chemical Identifiers

7440-48-4
58.933
GUTLYIVDDKVIGB-UHFFFAOYSA-N
104730
cobalt
Co
MFCD00010935
atom, co1+, cobalt, ion co1+, cobalto, cobaltum, hydride, hydrido, kobalt, moncation, powder
CHEBI:27638
[Co]
Safety and Handling

Safety and Handling

P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c

H317-H334-H350

EINECSNumber : 231-158-0

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents