Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt, Oil based standard solution, Specpure™, Co 1000μg/g

Chemical Identifiers

Chemical Name or Material Cobalt
Name Note Oil based standard solution
Purity Grade Specpure™
MDL Number MFCD00010935
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4714318

  SDS
alfa aesar™
4714318
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Cobalt
Quantity 50g
Name Note Oil based standard solution
Purity Grade Specpure™
Physical Form Liquid
Concentration Co 1000µg/g
MDL Number MFCD00010935