Please login to your online account to display your discounted pricing

Cobalt Nitrate, 8% (w/v), LabChem™

$60.29 - $60.29

Chemical Identifiers

Chemical Name or Material Cobalt Nitrate
CAS 7732-18-5
CAS Max % 92
CAS 10026-22-9
CAS Max % 8
Linear Formula Co(NO3)2·6H2O
Formula Weight 291.03
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC132207

 
lab chem
LC132207
125mL Each for $60.29
Description & Specifications

Specifications

Formula Weight 291.03
Chemical Name or Material Cobalt Nitrate
Identification Passes Test
Packaging Poly Bottle
Physical Form Liquid
Assay 8% (w/v)
Solubility Soluble in water
Storage Information Room Temperature
Linear Formula Co(NO3)2·6H2O
CAS 7732-18-5
Decomposition Information Nitrogen oxides; Cobalt and compounds
Color Clear pink
Quantity 125mL
Health Hazard 1 Danger
CAS Max % 92
Health Hazard 2 GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Toxic to aquatic life with long lasting effects.
CAS 10026-22-9
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If experiencing respiratory symptoms: Call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 8
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid breathing mist, vapors, spray.
Wear protective gloves, eye protection.
Wear respiratory protection.
Wash exposed skin thoroughly after handling.
Contaminated work clothing must not be allowed out of the workplace.
Avoid release to the environment.
If exposed or concerned: Get medical advice/attention.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash it before reuse.
If skin irritation or rash occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
If experiencing respiratory symptoms: Call a poison center/doctor.
Collect spillage.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May cause cancer.
May damage fertility or the unborn child.
Toxic to aquatic life with long lasting effects.