Please login to your online account to display your discounted pricing

Cobalt Nitrate, 6-Hydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$672.59 - $1,258.92

Chemical Identifiers

Chemical Name or Material Cobalt Nitrate
CAS 10026-22-9
Name Note 6-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula Co(NO3)2·6H2O
Formula Weight 291.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002629

 
avantor performance
168005
Poly Bottle 2.5kg Each for $1,258.92

02002630

 
avantor performance
168001
Poly Bottle 500g Each for $672.59
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 291.03
Chemical Name or Material Cobalt Nitrate
Molecular Formula Co(NO3)2·6H2O
CAS 10026-22-9
Name Note 6-Hydrate
Packaging Poly Bottle
Specific Gravity 1L = 1.87kg
Quantity 2.5kg