missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt nanopowder, APS 25-30nm, 99.8% (metals basis)

Manufacturer:  Alfa Aesar™ 4634714

 View more versions of this product

Catalog No. AA4634714



Specifications

Specifications

Cobalt nanopowder
7440-48-4
(metals basis)
MFCD00010935
GUTLYIVDDKVIGB-UHFFFAOYSA-N
[Co]
58.933
CHEBI:27638
Powder
25g
99.8%
Co
UN3089
Odorless
cobalt
104730
APS 25 to 30nm, Co3O4 layer ≈2nm

Chemical Identifiers

7440-48-4
58.933
GUTLYIVDDKVIGB-UHFFFAOYSA-N
CHEBI:27638
[Co]
Co
MFCD00010935
104730
cobalt
Safety and Handling

Safety and Handling

GHS H Statement
H228-H334-H351-H317
Flammable solid.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P210-P284a-P261-P342+P311-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

RTECSNumber : GF8750000

TSCA : TSCA

SDS Documents