missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(IV) sulfide, 99.5% (metals basis excluding Ni), Ni <0.2%

Manufacturer:  Alfa Aesar™ 1400506

 View more versions of this product

Catalog No. AA1400506



Specifications

Specifications

Cobalt(IV) sulfide
(metals basis excluding Ni), Ni <0.2%
MFCD00064735
cobalt 3+ ;trisulfide, cobalt sulfide,
[S-2].[S-2].[S-2].[Co+3].[Co+3]
214.046
123.06
99.5%
Odorless
5g
12013-10-4
Co2S3
UN3077
PZMYRTGXPBFNBW-UHFFFAOYSA-N
cobalt(3+);trisulfide
25147475
Powder
−200 Mesh
Black

Chemical Identifiers

12013-10-4
214.046
PZMYRTGXPBFNBW-UHFFFAOYSA-N
25147475
[S-2].[S-2].[S-2].[Co+3].[Co+3]
Co2S3
MFCD00064735
cobalt 3+ ;trisulfide, cobalt sulfide,
cobalt(3+);trisulfide
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : GG3310000

TSCA : TSCA

Documents