Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt iron oxide, may contain small amounts of iron oxide

Chemical Identifiers

Chemical Name or Material Cobalt iron oxide
CAS 12052-28-7
Name Note May contain small amounts of iron oxide
Formula Weight 234.62
MDL Number MFCD00016018
Synonym Cobalt ferrite
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2314706

 
alfa aesar™
2314706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2314714

 
alfa aesar™
2314714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 234.62
Chemical Name or Material Cobalt iron oxide
TSCA TSCA
Name Note May contain small amounts of iron oxide
Physical Form Powder
MDL Number MFCD00016018
CAS 12052-28-7
Synonym Cobalt ferrite
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H317
Toxic if swallowed.
May cause an allergic skin reaction.