Learn More
Cobalt(III) fluoride, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01149014
Description
Cobalt(III) fluoride is a powerful fluorinating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cobalt(III) fluoride | |
10026-18-3 | |
25 g | |
CoF3 | |
Insoluble in water. | |
Acrid | |
Light brown powder |
Brown | |
Powder | |
99% | |
MFCD00010942 | |
115.93 | |
Moisture sensitive | |
3.88 |
Chemical Identifiers
10026-18-3 | |
CoF3 | |
MFCD00010942 |
Safety and Handling
P210-P220-P221-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H272-H302+H312+H332-H314
DOTInformation : DOT Class: 5.1, Packaging Group: III
EINECSNumber : 233-062-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only