missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(III) 2,4-pentanedionate

Manufacturer:  Alfa Aesar™ 1253606

 View more versions of this product

Catalog No. AA1253606



Specifications

Specifications

Cobalt(III) 2,4-pentanedionate
Moisture Sensitive & Hygroscopic
5g
C15H21CoO6
Cobalt(III) acetylacetonate
356.26
Powder
Light
211°C (decomposition)
21679-46-9
MFCD00013488
VACIXPOMJJIDBM-UHFFFAOYSA-N
356.26

Chemical Identifiers

21679-46-9
356.26
VACIXPOMJJIDBM-UHFFFAOYSA-N
C15H21CoO6
MFCD00013488
Cobalt(III) acetylacetonate
Safety and Handling

Safety and Handling

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents