missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt(II) Tetraphenylporphyrin, TCI America™

Manufacturer:  TCI America C29861G

 View more versions of this product

Catalog No. C29861G



Specifications

Specifications

Cobalt(II) Tetraphenylporphyrin
1g
C44H28CoN4
5,10,15,20-Tetraphenyl-21H,23H-porphine Cobalt(II), Cobalt(II) TPP
C1=CC=C(C=C1)C2=C3C=CC(=C(C4=NC(=C(C5=CC=C([N-]5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C=C4)C9=CC=CC=C9)[N-]3.[Co+2]
671.669
671.67
≥98.0% (HPLC)
Undesignated
14172-90-8
MFCD00010725
LSZLYWSRWXFMOI-UHFFFAOYSA-N
cobalt(2+);5,10,15,20-tetraphenylporphyrin-22,24-diide
3994434
Crystal-Powder at 20°C

Chemical Identifiers

14172-90-8
671.669
LSZLYWSRWXFMOI-UHFFFAOYSA-N
3994434
C1=CC=C(C=C1)C2=C3C=CC(=C(C4=NC(=C(C5=CC=C([N-]5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)C=C4)C9=CC=CC=C9)[N-]3.[Co+2]
C44H28CoN4
MFCD00010725
5,10,15,20-Tetraphenyl-21H,23H-porphine Cobalt(II), Cobalt(II) TPP
cobalt(2+);5,10,15,20-tetraphenylporphyrin-22,24-diide
Safety and Handling

Safety and Handling

TSCA : No

Documents