missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) tetrafluoroborate hydrate, 96%

Manufacturer:  Alfa Aesar™ 2314222

 View more versions of this product

Catalog No. AA2314222



Specifications

Specifications

Cobalt(II) tetrafluoroborate hydrate
MFCD00016012
232.54 (Anhydrous)
96%
100g
26490-63-1
UN3260
Crystalline
Hygroscopic

Chemical Identifiers

26490-63-1
MFCD00016012
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H341-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents