missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt(II) sulfide, 99.5% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1311414

 View more versions of this product

Catalog No. AA1311414



Specifications

Specifications

Cobalt(II) sulfide
99.5% (Metal basis)
MFCD00016032
cobalt sulfide, sycoporite, cobalt ii sulfide, cobalt sulphide, cobaltous sulfide, cobalt sulfide, cobalt 2+ sulfide, cobalt monosulfide, cobalt sulfide amorphous, ccris 7728
S=[Co]
90.993
91
Odorless
25g
1317-42-6
CoS
Red powder
VRRFSFYSLSPWQY-UHFFFAOYSA-N
sulfanylidenecobalt
14832
Powder
Red

Chemical Identifiers

1317-42-6
90.993
VRRFSFYSLSPWQY-UHFFFAOYSA-N
14832
S=[Co]
CoS
MFCD00016032
cobalt sulfide, sycoporite, cobalt ii sulfide, cobalt sulphide, cobaltous sulfide, cobalt sulfide, cobalt 2+ sulfide, cobalt monosulfide, cobalt sulfide amorphous, ccris 7728
sulfanylidenecobalt
Safety and Handling

Safety and Handling

H400-H410-H317; Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. May cause an allergic skin reaction.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 215-273-3

Documents