missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt(II) sulfate hydrate, 99.999%, (trace metal basis), ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC379600500

 View more versions of this product

Catalog No. AC379600500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

155
50g
Trace metal basis
0.001% max.
Cobalt(II) sulfate hydrate
Glass bottle
60459-08-7
Crystalline Powder
Bordeaux
Co(II)SO4·xH2O
CoO4S·xH2O
99.999%
MFCD00149657

Chemical Identifiers

60459-08-7
CoO4S·xH2O
155
Cobalt(II) sulfate hydrate
99.999%
Co(II)SO4·xH2O
MFCD00149657
Trace metal basis

Safety and Handling

SDS & Safety Documents

GHS H Statement
Very toxic to aquatic life with long lasting effects. May cause cancer by inhalation. Harmful if swallowed. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Suspected of causing genetic defects. May damage fertility.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. Wear eye protection/face protection. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

Danger