Please login to your online account to display your discounted pricing

Cobalt(II) sulfate hydrate, 99.999%, (trace metal basis), ACROS Organics™

$138.37 - $138.37

Chemical Identifiers

Chemical Name or Material Cobalt(II) sulfate hydrate
CAS 60459-08-7
Purity Grade Trace metal basis
Assay Percent Range 99.999%
Molecular Formula CoO4S·xH2O
Linear Formula Co(II)SO4·xH2O
Formula Weight 155
MDL Number MFCD00149657
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC379600500

  SDS
thermo fisher scientific
AC379600500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC379600100

  SDS
thermo fisher scientific
AC379600100
Glass bottle 10g Each for $138.37
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 155
Quantity 50g
Color Bordeaux
Purity Grade Trace metal basis
Chemical Name or Material Cobalt(II) sulfate hydrate
Packaging Glass bottle
Assay Percent Range 99.999%
Molecular Formula CoO4S·xH2O
Linear Formula Co(II)SO4·xH2O
MDL Number MFCD00149657
Total Trace Metal Impurities 0.001% max.
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause cancer by inhalation.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defect
CAS 60459-08-7
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For ultra trace analysis

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
May cause cancer by inhalation.
Harmful if swallowed.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defect