Learn More
Cobalt(II) sulfate heptahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 1069622
Description
Cobalt(II) sulfate heptahydrate is used in the preparation of pigments and other cobalt salts. It is used in storage batteries and electroplating baths, sympathetic inks, and as an additive to soils and animal feeds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cobalt(II) sulfate heptahydrate | |
Red | |
Crystalline Aggregates | |
(metals basis) | |
CoSO4·7H2O | |
UN3077 | |
cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o | |
MEYVLGVRTYSQHI-UHFFFAOYSA-L | |
λ²-cobalt(2+) heptahydrate sulfate | |
61444 | |
99.999% | |
Odorless |
10026-24-1 | |
4 | |
100 g | |
CoH14O11S | |
MFCD00149658 | |
14,2448 | |
Soluble in water. | |
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O | |
281.09 | |
281.10 (154.99 Anhydrous) | |
Puratronic™ | |
2.03 g/mL |
Chemical Identifiers
10026-24-1 | |
281.09 | |
MEYVLGVRTYSQHI-UHFFFAOYSA-L | |
61444 |
CoH14O11S | |
MFCD00149658 | |
cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o | |
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O |
Safety and Handling
GHS H Statement
H334-H350-H360-H341-H302-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c
H302-H317-H334-H341-H350i-H360F
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 233-334-2
RTECSNumber : GG3200000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only