missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) sulfate heptahydrate, for analysis

CAS: 10026-24-1 | CoH14O11S | 281.09 g/mol

Supplier:  Thermo Scientific Chemicals 453900010

 View more versions of this product

Catalog No. AC453900010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cobalt(II) sulfate heptahydrate
10026-24-1
1 kg
CoSO4·7H2O
15,2432
Solubility in water: 362g/L (20°C). Other solubilities: slightly soluble in methanol and ethanol
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
281.09
281.09
Analytical
98.0°C
4 to 6 (5% aq. soln.)
CoH14O11S
MFCD00149658
cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o
MEYVLGVRTYSQHI-UHFFFAOYSA-L
λ²-cobalt(2+) heptahydrate sulfate
61444
99.0 to 101.0% (Complexometry)

Chemical Identifiers

10026-24-1
281.09
MEYVLGVRTYSQHI-UHFFFAOYSA-L
61444
CoH14O11S
MFCD00149658
cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENT

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only