Learn More
Cobalt(II) sulfate heptahydrate, 99+%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 213101000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cobalt(II) sulfate heptahydrate | |
99+% | |
Red to Brown | |
100 g | |
CoH14O11S | |
MFCD00149658 | |
cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o | |
MEYVLGVRTYSQHI-UHFFFAOYSA-L | |
λ²-cobalt(2+) heptahydrate sulfate | |
61444 | |
≥99% | |
Glass bottle |
98.0°C | |
10026-24-1 | |
Crystalline Powder and/or Crystals | |
99% min. (Complexometry) | |
CoSO4·7H2O | |
15,2432 | |
Solubility in water: 362g/L (20°C). Other solubilities: slightly soluble in methanol and ethanol | |
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O | |
281.09 | |
281.09 | |
Extra Pure | |
2.03 |
Chemical Identifiers
10026-24-1 | |
281.09 | |
MEYVLGVRTYSQHI-UHFFFAOYSA-L | |
61444 |
CoH14O11S | |
MFCD00149658 | |
cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o | |
O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
May
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENT
GHS Signal Word: Danger
RTECSNumber : GG3200000
RUO – Research Use Only