Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) stearate, Co 9-10%

$68.50 - $503.34

Chemical Identifiers

Chemical Name or Material Cobalt (II) stearate
CAS 1002-88-6
Molecular Formula C36H70CoO4
Linear Formula (C18H35O2)2Co
Formula Weight 625.89
MDL Number MFCD00054343
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4335230

 
alfa aesar™
4335230
250g Each for $68.50

AA43352A1

 
alfa aesar™
43352A1
1kg Each for $173.93

AA43352A7

 
alfa aesar™
43352A7
5kg Each for $503.34
Description & Specifications

Specifications

Physical Form Pellet
Formula Weight 625.89
Chemical Name or Material Cobalt (II) stearate
TSCA TSCA
Molecular Formula C36H70CoO4
Linear Formula (C18H35O2)2Co
MDL Number MFCD00054343
CAS 1002-88-6
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 1.71g/mL
RTECS Number RG1212000
Concentration Co 9 to 10%
Color Purple
Quantity 250g
Safety and Handling

Danger

GHS P Statement
P284a-P261-P280-P342+P311-P363-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.