missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) phosphate octahydrate

Manufacturer:  Alfa Aesar™ 2311118

 View more versions of this product

Catalog No. AA2311118



Specifications

Specifications

Cobalt(II) phosphate octahydrate
Co3H16O16P2
QOQSIXJUYVUEMP-UHFFFAOYSA-H
cobalt(2+);diphosphate;octahydrate
202398
Powder
Odorless
10294-50-5
MFCD00016028
O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]
510.859
510.87 (366.74 Anhydrous)
2.769g/mL
50g

Chemical Identifiers

10294-50-5
510.859
QOQSIXJUYVUEMP-UHFFFAOYSA-H
cobalt(2+);diphosphate;octahydrate
Co3H16O16P2
MFCD00016028
202398
O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]
Safety and Handling

Safety and Handling

GHS H Statement
H301-H341-H317
Toxic if swallowed.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P281-P301+P310-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : TB9279500

TSCA : TSCA

SDS Documents