missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cobalt(II) phosphate octahydrate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 02311130

 View more versions of this product

Catalog No. AA2311130



Description

Description

It is used as a pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Cobalt(II) phosphate octahydrate
Powder
Co3H16O16P2
cobalt ii phosphate octahydrate, cobaltous phosphate octahydrate, cobalt phosphate octahydrate, phosphoric acid, cobalt 2+ salt 2:3 , octahydrate, acmc-20ale3, 3co.2po4.8h2o, cobalt 2+ diphosphate octahydrate
QOQSIXJUYVUEMP-UHFFFAOYSA-H
cobalt(2+);diphosphate;octahydrate
202398
Odorless
10294-50-5
250 g
MFCD00016028
Slightly soluble in water. Soluble in mineral acids.
O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]
510.859
510.87 (366.74 Anhydrous)
2.769 g/mL

Chemical Identifiers

10294-50-5
510.859
QOQSIXJUYVUEMP-UHFFFAOYSA-H
202398
O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[O-]P(=O)([O-])[O-].[Co+2].[Co+2].[Co+2]
Co3H16O16P2
MFCD00016028
cobalt ii phosphate octahydrate, cobaltous phosphate octahydrate, cobalt phosphate octahydrate, phosphoric acid, cobalt 2+ salt 2:3 , octahydrate, acmc-20ale3, 3co.2po4.8h2o, cobalt 2+ diphosphate octahydrate
cobalt(2+);diphosphate;octahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H341-H317
Toxic if swallowed.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

P201-P202-P261-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c

H302-H315-H317-H319-H334-H335-H350

EINECSNumber : 236-655-6

RTECSNumber : TB9279500

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only