Alfa Aesar™ Cobalt(II) perchlorate hexahydrate, Reagent Grade
Manufacturer:  Alfa Aesar™ 1165230
 View more versions of this product
Catalog No. AA1165230
Specifications
Specifications
Cobalt(II) perchlorate hexahydrate
Cl2CoO8
UN1481
BSUSEPIPTZNHMN-UHFFFAOYSA-L
cobalt(2+);diperchlorate
26064
Crystalline
Odorless
250g
13478-33-6
MFCD00150207
cobaltous perchlorate, cobalt diperchlorate, cobalt ii perchlorate, cobalt diperchlorate hexahydrate, cobalt ii perchlorate, cobalt perchlorate hexahydrate, cobaltous perchlorate, hexahydrate, cobalt ii perchlorate, hexahydrate, perchloric acid, cobalt ii salt, hexahydrate, diperchloratocobalt
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2]
257.825
366.74 (257.83 Anhydrous)
Reagent
Hygroscopic
Chemical Identifiers
13478-33-6
257.825
BSUSEPIPTZNHMN-UHFFFAOYSA-L
26064
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2]
Cl2CoO8
MFCD00150207
cobaltous perchlorate, cobalt diperchlorate, cobalt ii perchlorate, cobalt diperchlorate hexahydrate, cobalt ii perchlorate, cobalt perchlorate hexahydrate, cobaltous perchlorate, hexahydrate, cobalt ii perchlorate, hexahydrate, perchloric acid, cobalt ii salt, hexahydrate, diperchloratocobalt
cobalt(2+);diperchlorate
Safety and Handling
Safety and Handling
GHS H Statement
H272-H351-H315-H319-H317-H335
May intensify fire
oxidizer.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
TSCA : TSCA
Documents