Alfa Aesar™ Cobalt(II) perchlorate hexahydrate, Reagent Grade
Manufacturer:  Alfa Aesar™ 1165218
 View more versions of this product
Catalog No. AA1165218
Specifications
Specifications
Cobalt(II) perchlorate hexahydrate
Cl2CoO8
UN1481
BSUSEPIPTZNHMN-UHFFFAOYSA-L
cobalt(2+);diperchlorate
26064
Crystalline
Odorless
50g
13478-33-6
MFCD00150207
cobalt diperchlorate
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2]
257.825
366.74 (257.83 Anhydrous)
Reagent
Hygroscopic
Chemical Identifiers
13478-33-6
257.825
BSUSEPIPTZNHMN-UHFFFAOYSA-L
26064
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2]
Cl2CoO8
MFCD00150207
cobalt diperchlorate
cobalt(2+);diperchlorate
Safety and Handling
Safety and Handling
GHS H Statement
H272-H351-H315-H319-H317-H335
May intensify fire
oxidizer.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.
TSCA : TSCA
Documents