missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) perchlorate hexahydrate, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4432009

 View more versions of this product

Catalog No. AA4432009



Specifications

Specifications

Cobalt (II) perchlorate hexahydrate
(metals basis)
MFCD00150207
cobaltous perchlorate, cobalt diperchlorate, cobalt ii perchlorate, cobalt diperchlorate hexahydrate, cobalt ii perchlorate, cobalt perchlorate hexahydrate, cobaltous perchlorate, hexahydrate, cobalt ii perchlorate, hexahydrate, perchloric acid, cobalt ii salt, hexahydrate, diperchloratocobalt
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2]
257.825
366.74 (257.83 Anhydrous)
99.999%
Hygroscopic
13478-33-6
Cl2CoO8
UN1481
BSUSEPIPTZNHMN-UHFFFAOYSA-L
cobalt(2+);diperchlorate
26064
Crystalline
Odorless
10g

Chemical Identifiers

13478-33-6
257.825
BSUSEPIPTZNHMN-UHFFFAOYSA-L
26064
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2]
Cl2CoO8
MFCD00150207
cobaltous perchlorate, cobalt diperchlorate, cobalt ii perchlorate, cobalt diperchlorate hexahydrate, cobalt ii perchlorate, cobalt perchlorate hexahydrate, cobaltous perchlorate, hexahydrate, cobalt ii perchlorate, hexahydrate, perchloric acid, cobalt ii salt, hexahydrate, diperchloratocobalt
cobalt(2+);diperchlorate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H351-H315-H319-H317-H335
May intensify fire
oxidizer.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents