missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) perchlorate hexahydrate, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 4432030

 View more versions of this product

Catalog No. AA4432030



Specifications

Specifications

Cobalt (II) perchlorate hexahydrate
(metals basis)
MFCD00150207
cobalt diperchlorate
BSUSEPIPTZNHMN-UHFFFAOYSA-L
cobalt(2+);diperchlorate
26064
Crystalline
Odorless
250g
13478-33-6
Cl2CoO8
UN1481
Soluble in water.
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2]
257.825
366.74 (257.83 Anhydrous)
99.999%
Hygroscopic

Chemical Identifiers

13478-33-6
257.825
BSUSEPIPTZNHMN-UHFFFAOYSA-L
26064
[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2]
Cl2CoO8
MFCD00150207
cobalt diperchlorate
cobalt(2+);diperchlorate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H351-H315-H319-H317-H335
May intensify fire
oxidizer.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P201-P202-P210-P220-P221-P261-P264b-P271-P280i-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P342+P311-P362-P370+P378q-P501c

H272-H315-H319-H334-H335-H350

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, N.O.S.

EINECSNumber : 236-653-5

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents