Learn More
Cobalt(II) oxide, Co >74%, -325 mesh, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 379200100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cobalt(II) oxide | |
Brown-Green or Gray | |
Powder | |
99+% | |
MFCD00016031 | |
cobalt ii oxide, zaffre, cobalt monoxide, cobalt black, cobalt oxide, cobaltous oxide, cobalt monooxide, monocobalt oxide, cobalt 2+ oxide, ci pigment black 13 | |
IVMYJDGYRUAWML-UHFFFAOYSA-N | |
oxocobalt | |
14786 | |
>74% (Co) |
1785.0°C | |
1307-96-6 | |
10 g | |
CoO | |
15,243 | |
Solubility in water: insoluble | |
O=[Co] | |
74.93 | |
74.93 | |
Glass bottle |
Chemical Identifiers
1307-96-6 | |
74.93 | |
IVMYJDGYRUAWML-UHFFFAOYSA-N | |
14786 | |
O=[Co] |
CoO | |
MFCD00016031 | |
cobalt ii oxide, zaffre, cobalt monoxide, cobalt black, cobalt oxide, cobaltous oxide, cobalt monooxide, monocobalt oxide, cobalt 2+ oxide, ci pigment black 13 | |
oxocobalt |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Fatal if inhaled.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF INHALED: Remove to fresh air and keep at rest in a posit
GHS Signal Word: Danger
EINECSNumber : 215-154-6
RUO – Research Use Only