missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) oxalate dihydrate, Reagent Grade

Stabilizer for HCN, temperature indicator

Manufacturer:  Alfa Aesar™ 8775806

 View more versions of this product

Catalog No. AA8775806



Specifications

Specifications

Cobalt (II) oxalate dihydrate
Odorless
5g
Reagent
MFCD00050793
14,2445
Soluble in aqueous ammonia; Insoluble in water; Slightly soluble in acids
C(=O)(C(=O)[O-])[O-].O.O.[Co+2]
182.981
182.98 (146.95 Anhydrous)
3.021g/mL
229°C to 231°C (decomposition)
5965-38-8
C2H4CoO6
UN3288
cobaltousoxalate,cobaltoxalate
MWHSMSAKVHVSAS-UHFFFAOYSA-L
cobalt(2+);oxalate;dihydrate
131881712
Powder

Chemical Identifiers

5965-38-8
182.981
MWHSMSAKVHVSAS-UHFFFAOYSA-L
131881712
C(=O)(C(=O)[O-])[O-].O.O.[Co+2]
C2H4CoO6
MFCD00050793
cobaltousoxalate,cobaltoxalate
cobalt(2+);oxalate;dihydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents