missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) nitrate hexahydrate, Puratronic™, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1069409

 View more versions of this product

Catalog No. AA1069409



Specifications

Specifications

Cobalt(II) nitrate hexahydrate
(metals basis)
MFCD00149647
14,2444
QGUAJWGNOXCYJF-UHFFFAOYSA-N
cobalt(2+);dinitrate;hexahydrate
24821
291.04 (182.94 Anhydrous)
99.999%
1.87g/mL
1.52
55°C to 56°C
10026-22-9
CoH12N2O12
UN1477
cobalt 2+ nitrate hexahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Co+2]
291.031
CHEBI:86214
Wet Crystalline Aggregates
Puratronic™
Odorless
Red
10g

Chemical Identifiers

10026-22-9
291.031
QGUAJWGNOXCYJF-UHFFFAOYSA-N
24821
cobalt(2+);dinitrate;hexahydrate
CoH12N2O12
MFCD00149647
cobalt 2+ nitrate hexahydrate
CHEBI:86214
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Co+2]
Safety and Handling

Safety and Handling

GHS H Statement
H272-H334-H350-H360-H341-H302-H317
May intensify fire; oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
P221-P210-P284a-P342+P311-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

RTECSNumber : QU7355500

TSCA : TSCA

Documents