Learn More
Cobalt(II) nitrate hexahydrate, 97.7% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01134114
Description
Cobalt(II) nitrate hexahydrate is used in the preparation of metallic cobalt and cobalt based dyes and pigments. It is a precursor for the preparation of catalyst for use in certain organic reactions. It finds use in catalysis, vitamin preparations, and porcelain decoration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cobalt(II) nitrate hexahydrate | |
10026-22-9 | |
Crystalline | |
CoH12N2O12 | |
UN1477 | |
cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate | |
QGUAJWGNOXCYJF-UHFFFAOYSA-N | |
cobalt(2+);dinitrate;hexahydrate | |
24821 | |
291.04 (182.94 Anhydrous) | |
Odorless | |
1.87 g/mL |
55°C to 56°C | |
Red | |
25 g | |
MFCD00149647 | |
14,2444 | |
Soluble in water,alcohol,acetone and ammonia. | |
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
291.03 | |
CHEBI:86214 | |
≤97.7% | |
1.52 |
Chemical Identifiers
10026-22-9 | |
291.03 | |
QGUAJWGNOXCYJF-UHFFFAOYSA-N | |
24821 | |
cobalt(2+);dinitrate;hexahydrate |
CoH12N2O12 | |
MFCD00149647 | |
cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate | |
CHEBI:86214 | |
O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Safety and Handling
GHS H Statement
H272-H334-H350-H360-H341-H302-H317
May intensify fire; oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Suspected of causing genetic defects.
Harmful if swallowed.
May cause an allergic skin reaction.
P201-P202-P210-P220-P221-P261-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P330-P333+P313-P342+P311-P363-P370+P378q-P501c
H272-H302+H332-H317-H318-H334-H341-H350i-H360F
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber : 233-402-1
RTECSNumber : QU7355500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only