Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) meso-tetrakis(4-methoxyphenyl)porphine

$82.66 - $253.99

Chemical Identifiers

Chemical Name or Material Cobalt(II) meso-tetrakis(4-methoxyphenyl)porphine
CAS 28903-71-1
Molecular Formula C48H36CoN4O4
Formula Weight 791.79
MDL Number MFCD00010724
Synonym 5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine cobalt(II)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2594503

  SDS
alfa aesar™
H2594503
1g Each for $82.66

AAH2594506

  SDS
alfa aesar™
H2594506
5g Each for $253.99
Description & Specifications

Specifications

Formula Weight 791.79
Chemical Name or Material Cobalt(II) meso-tetrakis(4-methoxyphenyl)porphine
Molecular Formula C48H36CoN4O4
MDL Number MFCD00010724
CAS 28903-71-1
Synonym 5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine cobalt(II)
Melting Point >300°C
Quantity 1g