Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) iodide, ultra dry, 99.999% (metals basis)

Chemical Identifiers

Chemical Name or Material Cobalt(II) iodide
CAS 15238-00-3
Name Note Ultra dry
Formula Weight 312.74
MDL Number MFCD00016030
Merck Index 14,2443
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3573003

  SDS
alfa aesar™
3573003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3573006

  SDS
alfa aesar™
3573006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 515°C (vac.)
Boiling Point 570°C (vac.)
Formula Weight 312.74
Packaging Ampule
Sensitivity Light Sensitive & Hygroscopic
Physical Form Beads
Storage Note 1
  • Ampouled under argon
Assay 99.999%
Name Note Ultra dry
Assay Percent Notes (metals basis)
CAS 15238-00-3
Density 5.584g/mL
Quantity 1g
Odor Odorless
Merck Index 14,2443
Health Hazard 2 GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
TSCA TSCA
Health Hazard 3 GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color White
Chemical Name or Material Cobalt(II) iodide
MDL Number MFCD00016030
Health Hazard 1 Warning
Mesh Size −10 Mesh

  • Ampouled under argon

Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.

  • Ampouled under argon