Learn More
Cobalt(II) iodide, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals
Moisture and humidity indicator, catalyst
Supplier: Thermo Scientific Chemicals 02311922
Description
Cobalt(II) iodide is used as a moisture and humidity indicator. It is also used as a catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
570°C (vac.) | |
515°C (vac.) | |
15238-00-3 | |
100 g | |
MFCD00016030 | |
cobalt ii iodide, acmc-20ale6, cobalt ii iodide, anhydrous, cobalt ii iodide, ultra dry, cobalt ii iodide, powder, cobalt ii iodide, anhydrous, powder trace metals basis | |
312.74 | |
Odorless |
Cobalt(II) iodide | |
Anhydrous | |
Powder | |
(metals basis) | |
14,2443 | |
Soluble in water | |
99.5% | |
Hygroscopic; Light sensitive |
Chemical Identifiers
15238-00-3 | |
MFCD00016030 | |
cobalt ii iodide, acmc-20ale6, cobalt ii iodide, anhydrous, cobalt ii iodide, ultra dry, cobalt ii iodide, powder, cobalt ii iodide, anhydrous, powder trace metals basis |
Safety and Handling
GHS H Statement
H341-H317
Suspected of causing genetic defects.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H341-H350
EINECSNumber : 239-283-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only