Please login to your online account to display your discounted pricing

Cobalt(II,III) oxide, 99% (metals basis), Alfa Aesar™

S.A. 40-70m2/g

Chemical Identifiers

Chemical Name or Material Cobalt(II, III) oxide
CAS 1308-06-1
Physical Form Powder
Assay Percent Range 99% (Metal basis)
Molecular Formula Co3O4
Formula Weight 240.8g/mol
MDL Number MFCD00010939
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4466014

 
alfa aesar
44660-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Appearance Black powder
Density 6.11g/cm3
Odor Odorless
Color Black
Quantity 25g
Chemical Name or Material Cobalt(II, III) oxide
CAS 1308-06-1
Physical Form Powder
Assay Percent Range 99% (Metal basis)
Molecular Formula Co3O4
Formula Weight 240.8g/mol
MDL Number MFCD00010939
Health Hazard 1 H351-H400-H410-H317; Suspected of causing cancer. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. May cause an allergic skin reaction.
DOT Information DOT Class: 9, Packaging Group: III
EINECS Number 215-157-2
Safety and Handling

H351-H400-H410-H317; Suspected of causing cancer. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects. May cause an allergic skin reaction.