Learn More
Cobalt(II) hydroxide, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 012524A3
Description
Cobalt(II) hydroxide is used as a drying agent for paints and varnishes. It acts as an aid to dry the lithographic inks and catalyst in the preparation of battery electrode impregnating solutions. It is also involved in the synthesis of other cobalt compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Cobalt(II) hydroxide | |
21041-93-0 | |
Powder | |
(metals basis) | |
Co(OH)2 | |
14,2442 | |
Soluble in acids and ammonia; Very slightly soluble in water; Insoluble in dilute alkalis. | |
[OH-].[OH-].[Co++] | |
92.95 | |
92.95 | |
Odorless |
250°C (decomposition) | |
Pink to Purple | |
2 kg | |
CoH2O2 | |
MFCD00016015 | |
cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis | |
ASKVAEGIVYSGNY-UHFFFAOYSA-L | |
cobalt(2+);dihydroxide | |
10129900 | |
99.9% | |
Air Sensitive |
Chemical Identifiers
21041-93-0 | |
92.95 | |
ASKVAEGIVYSGNY-UHFFFAOYSA-L | |
10129900 | |
[OH-].[OH-].[Co++] |
CoH2O2 | |
MFCD00016015 | |
cobalt hydroxide, cobalt ii hydroxide, cobaltous hydrate, acmc-20aleg, cobalt 2+ dihydroxide, cobalt ii hydroxide metals basis | |
cobalt(2+);dihydroxide |
Safety and Handling
GHS H Statement
H302-H317
Harmful if swallowed.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P363-P501c
H302+H332-H317-H319-H334-H350
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 244-166-4
RTECSNumber : GG0904500
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only