Please login to your online account to display your discounted pricing

Alfa Aesar™ Cobalt(II) fluoride, ultra dry, 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Cobalt(II) fluoride
CAS 10026-17-2
Name Note Ultra dry
Formula Weight 96.93
MDL Number MFCD00010941
UN Number UN2923
Merck Index 14,2440
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3574004

  SDS
alfa aesar™
3574004
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3574009

  SDS
alfa aesar™
3574009
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3574018

  SDS
alfa aesar™
3574018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 96.93
Packaging Ampule
Assay 99.99%
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
CAS 10026-17-2
Health Hazard 2 GHS H Statement
H301-H314-H351-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause an allergic skin reaction.
Chemical Name or Material Cobalt(II) fluoride
Quantity 2g
Assay Percent Notes (metals basis)
MDL Number MFCD00010941
Storage Note 1
  • Ampouled under argon
Melting Point 1200°C
Health Hazard 1 Danger
Odor Odorless
RTECS Number GG0770000
UN Number UN2923
Boiling Point 1400°C
Density 4.46g/mL
TSCA TSCA
Sensitivity Hygroscopic
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Powder
Name Note Ultra dry
Merck Index 14,2440

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H351-H317
Toxic if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing cancer.
May cause an allergic skin reaction.

  • Ampouled under argon